Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STAT3, Clone: 21E7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP148014

Catalog No. NBP148014


Add to cart

Description

Description

STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

STAT3
OTI21E7
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Mouse
IgG1
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens signal transducer and activator of transcription 3 (acute-phase response factor) (STAT3), transcript variant 3.
Immunoassay, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Knockout Validated
Affinity Purified
STAT3
Recombinant protein expressed in E.coli corresponding to amino acids 154-175 of human stat3
88.08 kDa
0.1 ml
Cellular Markers, Signal Transduction, Transcription Factors and Regulators
Primary
6774
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only