Learn More
anti-STAT5a (p Tyr694), DyLight 488, Clone: 5F6.F1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP177817G
Catalog No. NBP177817G
Add to cart
Description
Description
STAT5A p Tyr694 Monoclonal antibody specifically detects STAT5A p Tyr694 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
STAT5a (p Tyr694)
5F6.F1
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
STAT5A
This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5a protein.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody is specific for mouse Stat5a protein phosphorylated at Y694. A BLAST analysis was used to suggest cross-reactivity with Stat5a from human, mouse.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
MGF, signal transducer and activator of transcription 5A, STAT5
Mouse
IgG1 Kappa
0.1 ml
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
6776
Human, Mouse, Rat