Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-STAT5b, Clone: 5B3, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP151630

Catalog No. NBP151630

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

STAT5b
5B3
Western Blot 1:200-1:1000, ELISA 1:10000
Ascites
STAT5B
Purified recombinant fragment of human STAT5B expressed in E. Coli.
90 kDa
0.1 ml
Signal Transduction
Primary
6777
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only