Learn More
anti-STRO-1, Alexa Fluor 405, Clone: STRO-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP148356AF405
Catalog No. NBP148356W
Add to cart
Description
Description
STRO-1 Monoclonal antibody specifically detects STRO-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
STRO-1
STRO-1
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgM Lambda
0.1 ml
Store at 4C in the dark.
Monoclonal
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
STRO1
Human CD34+ bone marrow cells.
IgM purified
RUO
Primary
Human