Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Sulfatase-2/SULF2, APC, Clone: 2B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP136727APC

Catalog No. NBP136727J


Add to cart

Description

Description

Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Sulfatase-2/SULF2
2B4
Flow Cytometry
Purified
SULF2
Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998].
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Does not recognize human or mouse Sulfatase 1 or other family members.
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2
Mouse
IgG1
0.1 ml
Lipid and Metabolism
Primary
55959
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only