Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Sulfatase-2/SULF2, DyLight 405, Clone: 2B4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP136727V
Description
Sulfatase-2/SULF2 Monoclonal antibody specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Sulfatase-2/SULF2 | |
2B4 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
NP_940998 | |
DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
Mouse | |
IgG1 | |
0.1 ml | |
Lipid and Metabolism | |
Primary | |
55959 | |
Human, Mouse |
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified | |
SULF2 | |
Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998]. | |
Protein G purified | |
RUO | |
Store at 4C in the dark. | |
Monoclonal | |
Does not recognize human or mouse Sulfatase 1 or other family members. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only