Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SUZ12, Clone: 3D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151687

Catalog No. NBP151687


Add to cart

Description

Description

SUZ12 Monoclonal antibody specifically detects SUZ12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SUZ12
3D10
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Ascites
SUZ12
Purified recombinant fragment of human SUZ12 expressed in E. Coli.
83.1 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
ChET 9 protein, CHET9polycomb protein SUZ12, Chromatin precipitated E2F target 9 protein, JJAZ1Joined to JAZF1 protein, KIAA0160Suppressor of zeste 12 protein homolog, suppressor of zeste 12 homolog (Drosophila)
Mouse
IgG1
Unpurified
RUO
Primary
23512
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only