Learn More
anti-Syntaxin 1A, Clone: SP6, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP119331
Catalog No. NBP119331
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Syntaxin 1A
SP6
Western Blot 1:100-1:2000
Purified
STX1A
Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin mAb antibodies
Protein G purified
RUO
Primary
6804
Mouse, Rat
Western Blot
Unconjugated
No buffer with Sodium Azide
HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A
Mouse
IgG1
0.05 mg
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Syntaxin (SP6)