Learn More
anti-Synuclein-alpha, Clone: 3H9, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192694SS
 View more versions of this product
Catalog No. NBP192694SS
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
alpha-Synuclein
3H9
Unconjugated
No buffer with No Preservative
Ascites
SNCA
Amino acids 61-95 of human alpha-Synuclein, which correspond to the Non-Amyloid beta Component of Alzheimer's disease amyloid (NAC). [UniProt# P37840]
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes full length human and rodent alpha Synuclein. It will also bind human alpha Synuclein containing the A30P and A53T mutations.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
1.0 mg/mL
Western Blot, Simple Western 1:10, Flow Cytometry 1:50 - 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Paraffin 1:1000, CyTOF-ready
P37840
Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Mouse
IgG1
0.025 ml
Alzheimers Research, Apoptosis, Neurodegeneration, Neuroscience
Primary
6622
Human, Mouse, Rat