Learn More
anti-Synuclein-alpha (Nitrate Tyr125, Nitrate Tyr133), DyLight 405, Clone: 24.8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP126380V
Catalog No. NBP126380V
Add to cart
Description
Description
alpha-Synuclein Nitrate Tyr125, Nitrate Tyr133 Monoclonal antibody specifically detects alpha-Synuclein Nitrate Tyr125, Nitrate Tyr133 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
alpha-Synuclein (Nitrate Tyr125, Nitrate Tyr133)
24.8
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
P37840
Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SNCA
Nitrated human alpha-Synuclein [Uniprot: P37840]
14 kDa
0.1 ml
Alzheimers Research, Apoptosis, Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Stem Cell Markers
Primary
6622