Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-T-bet/TBX21, PerCP, Clone: 4B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143298PCP

Catalog No. NBP143298P


Add to cart

Description

Description

T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

T-bet/TBX21
4B10
Western Blot, Flow Cytometry, Immunoprecipitation
Affinity Purified
TBX21
The immunogen for this antibody was TBet.
Protein A or G purified
RUO
Primary
30009
Flow Cytometry, Immunoprecipitation, Western Blot
PerCP
PBS with 0.05% Sodium Azide
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rhesus Macaque
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only