Learn More
TAF15 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: 4D71, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192685V
Catalog No. NBP192685V
Add to cart
Description
Description
TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
TAF15
4D71
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
TAF15
Full length recombinant human TAF15 expressed in and purified from E. coli. [UniProt# Q92804]
68 kDa
0.1 ml
DNA Repair, Neuroscience
Primary
8148
Immunocytochemistry, Immunofluorescence, Western Blot
1mg/mL
Western Blot, Immunocytochemistry/Immunofluorescence
Q92804
68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat