Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TAG-72, APC, Clone: CC49, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233128APC

Catalog No. NBP233128J


Add to cart

Description

Description

TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine samples. It is validated for Flow Cytometry.
Specifications

Specifications

TAG-72
CC49
Flow Cytometry
Purified
Mouse
IgG1 Kappa
Protein A purified
RUO
Primary
182875
Human, Rat, Bovine, Canine
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72
Purified TAG-72 protein
220 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes an oncofetal antigen of 220kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only