Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TBX5, Clone: 7B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147450

Catalog No. NBP147450


Add to cart

Description

Description

TBX5 Monoclonal antibody specifically detects TBX5 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

TBX5
7B11
Western Blot 1: 500-1:2000, ELISA 1:10000
Ascites
TBX5
Purified recombinant fragment of TBX5 expressed in E. Coli.
53 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
HOS, T-box 5, T-box protein 5, T-box transcription factor TBX5
Mouse
IgG1
Unpurified
RUO
Primary
6910
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only