Learn More
anti-TCR alpha/beta, Biotin, Clone: TCR3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128285
Catalog No. NBP128285
Add to cart
Description
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Avian, Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
TCR alpha/beta
TCR3
Biotin
PBS with 0.05% Sodium Azide
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Mouse
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken TCR alpha beta (Vbeta2)
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro assay
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
TRA
CD3+ TCR1- TCR2- Ia- chicken blood mononuclear cells
Protein A or G purified
RUO
Primary
6955
Avian, Chicken