Learn More
anti-TCR gamma/delta, Biotin, Clone: TCR1, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128277
Catalog No. NBP128277
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
TCR gamma/delta
TCR1
Biotin
PBS with 0.05% Sodium Azide
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Mouse
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken, Reptile
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
TRG
Outbred chicken thymocytes and Ig-negative blood lymphocytes
Protein A or G purified
RUO
Primary
6965