Learn More
anti-TCR gamma/delta, DyLight 488, Clone: B1.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143121G
Catalog No. NBP143121G
Add to cart
Description
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TCR gamma/delta
B1.1
Flow Cytometry
Affinity Purified
TRG
The immunogen for this antibody was gamma delta TCR.
Protein A or G purified
RUO
Primary
6965
Flow Cytometry
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human