Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TDP1, Clone: 1F2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130357

Catalog No. NBP130357


Add to cart

Description

Description

TDP1 Monoclonal antibody specifically detects TDP1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

TDP1
1F2
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Purified
TDP1
Recombinant human TDP1 (1-298aa) purified from E. coli
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This TDP1 monoclonal antibody, clone 1F2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TDP1 protein.
ELISA, Western Blot
1.0 mg/mL
Western Blot 1:1000-1:2000, ELISA 1:100-1:2000
NP_060789
EC 3.1.4, EC 3.1.4.-, FLJ11090, MGC104252, SCAN1, Tyr-DNA phosphodiesterase 1, tyrosyl-DNA phosphodiesterase 1
Mouse
IgG2b Kappa
0.1 ml
Stem Cell Markers
Primary
55775
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only