Learn More
anti-Testosterone, Clone: P2G1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178589
Catalog No. NBP178589
Add to cart
Description
Description
Testosterone Monoclonal antibody specifically detects Testosterone in Non-species specific samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications
Specifications
Testosterone
P2G1
Unconjugated
10mM Potassium Phosphate, 2.5mM Potassium Chloride, 135mM NaCl and 1.0 mg/ml BSA with 0.05% Sodium Azide
Mouse
IgG1
100ug
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Other
ELISA, Immunoprecipitation, Radioimmune Assays
1 mg/mL
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Affinity Purified
Testosterone-3-CMO-BSA
Ion exchange chromatography
RUO
Primary
Chemical.