Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta 1 Mouse anti-Human, Rat, Mustelid, Sheep, Clone: TB21, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151749

Catalog No. NBP151749


Add to cart

Description

Description

TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human, Rat, Mustelid, Sheep samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

TGF-beta 1
TB21
Unconjugated
PBS with 0.09% Sodium Azide
Purified
TGFB1
The antibody reacts with the TGF beta 2.
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes both human platelet-derived and recombinant TGF-beta1 in enzymelinked immunosorbent assay (ELISA). Clone TB21 demonstrates neutralising activity against TGF-beta1 in cell proliferation assays. Removal of sodium azide is recommended prior to use
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
1.0 mg/mL
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10 ug/ml, Immunohistochemistry-Frozen 10 ug/ml
P61812
CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Mouse
IgG1
0.025 mg
Asthma, Cancer, Immunology
Primary
7040
Human, Mustelid, Rat, Sheep
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only