Learn More
anti-Thyroid receptor-interacting protein 11, Alexa Fluor 700, Clone: 10G5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141204AF700
Catalog No. NBP141204Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thyroid receptor-interacting protein 11 Monoclonal antibody specifically detects Thyroid receptor-interacting protein 11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Thyroid receptor-interacting protein 11
10G5
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
TRIP11
Amino acids 1099-1372 of Trip230 expressed in E. coli.
Protein G purified
RUO
Primary
9321
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CEV14TR-interacting protein 11, Clonal evolution-related gene on chromosome 14 protein, GMAP-210ACG1A, Golgi-associated microtubule-binding protein 210, Golgi-microtubule-associated protein of 210 kDa, thyroid hormone receptor interactor 11, thyroid receptor-interacting protein 11, TRIP-11, Trip230
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody will recognize human TRIP230.