Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Thyroid receptor-interacting protein 11, Clone: 10G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141204

Catalog No. NBP141204


Add to cart

Description

Description

Thyroid receptor-interacting protein 11 Monoclonal antibody specifically detects Thyroid receptor-interacting protein 11 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Thyroid receptor-interacting protein 11
10G5
Unconjugated
PBS (pH 7.4) with No Preservative
CEV14TR-interacting protein 11, Clonal evolution-related gene on chromosome 14 protein, GMAP-210ACG1A, Golgi-associated microtubule-binding protein 210, Golgi-microtubule-associated protein of 210 kDa, thyroid hormone receptor interactor 11, thyroid receptor-interacting protein 11, TRIP-11, Trip230
Mouse
IgG1
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody will recognize human TRIP230.
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1.0mg/mL
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Purified
TRIP11
Amino acids 1099-1372 of Trip230 expressed in E. coli.
Immunogen affinity purified
RUO
Primary
9321
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only