Learn More
anti-Thyroid Stimulating Hormone, Clone: Fc1002, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178580
Catalog No. NBP178580
Add to cart
Description
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications
Specifications
Thyroid Stimulating Hormone
Fc1002
Unconjugated
10mM Potassium Phosphate, 2.5mM Potassium Chloride, 135mM NaCl and 1.0 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
TSHB
Human Thyroid Stimulating Hormone
Ion exchange chromatography
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Immunoprecipitation, Radioimmune Assays
1 mg/mL
ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
P01215
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA
Mouse
IgG1
100ug
Cancer
Primary
7252