Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-TLR2, Clone: TL2.3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP151793
Catalog No. NBP151793
Description
Description
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay.
Specifications
Specifications
TLR2
TL2.3
Unconjugated
0.2 um filtered solution in PBS and 0.1% BSA with No Preservative
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Mouse
IgG2a
0.05 mg
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary
7097
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Immunoassay
Purified
TLR2
Human TLR2-expressing CHO cells
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human