Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1530
This item is not returnable.
View return policy
Description
TLR2 Polyclonal specifically detects TLR2 in Mouse samples. It is validated for Western Blot.Specifications
TLR2 | |
Polyclonal | |
Unconjugated | |
Q9QUN7 | |
TLR2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TLR2 Gln25-Leu590 Accession # Q9QUN7 | |
100 μg | |
Primary | |
Detects mouse TLR2 in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human (rh) TLR2 is observed and less than 2% cross-reactivity with recombinant mouse (rm) TLR1, rhTLR3, rmTLR4, and rmTLR6 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Goat | |
Affinity Purified | |
RUO | |
7097 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title