Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Antibody (85B152.5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197728B
Description
TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TLR5 | |
Monoclonal | |
Biotin | |
O60602 | |
TLR5 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259). | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Canine | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
85B152.5 | |
Western Blot, Immunohistochemistry-Paraffin | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
Mouse | |
100 kDa | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
7100 | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction