Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TLR9, Clone: 5G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151156

Catalog No. NBP151156


Add to cart

Description

Description

TLR9 Monoclonal antibody specifically detects TLR9 in Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

TLR9
5G5
Unconjugated
PBS, containing 0.1% BSA with 0.02% Sodium Azide
Purified
TLR9
Purified fusion protein of extracellular domain of human TLR9 (AA 1-815) and human IgGFc
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Canine, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
Western Blot 2 ug/ml, Flow Cytometry 1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 4ug/400ul, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:100
Q9NR96
CD289, CD289 antigen, toll-like receptor 9
Mouse
IgG2a
0.05 mg
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
54106
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only