Learn More
anti-Topoisomerase I, Clone: 1291CT875.142.166, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232310
Catalog No. NBP232310
Add to cart
Description
Description
Topoisomerase I Monoclonal antibody specifically detects Topoisomerase I in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Flow (Cell Surface).
Specifications
Specifications
Topoisomerase I
1291CT875.142.166
Unconjugated
PBS with 0.09% Sodium Azide
Purified
TOP1
This TOP1 antibody is generated from a mouse immunized with a recombinant protein from the N-terminal region of human TOP1.
90.726 kDa
400 ul
Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing
Primary
7150
Western Blot, Flow Cytometry
0.5 mg/mL
Western Blot 1:1000, Flow Cytometry 1:25, Flow (Cell Surface) 1:10-1:1000
P11387
DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase
Mouse
IgG1 Kappa
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Rat