Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRAIL R2/TNFRSF10B, Clone: DR5-01-1, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145024

Catalog No. NBP145024

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

TRAIL R2/TNFRSF10B
DR5-01-1
Unconjugated
PBS (pH 7.4) with 15mM Sodium Azide
Purified
TNFRSF10B
Recombinant fusion protein of human IgG heavy chain and extracellular domain of DR5 (NM_003842.4).
Protein A purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody DR5-01-1 recognizes an extracellular domain of TRAIL-R2 (DR5). TRAIL-R2 is one of two TNF superfamily member intracellular death domain containing receptors for TRAIL (APO2L).
Western Blot, Flow Cytometry, CyTOF
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1-10 ug/ml, CyTOF-ready
O14763
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TRAIL receptor 2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Mouse
IgG1
0.1 mg
Apoptosis, Cancer
Primary
8795
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only