Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Transglutaminase 4/TGM4, Clone: 1H1, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP128609

Catalog No. NBP128609

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Transglutaminase 4/TGM4
1H1
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
TGM4
Recombinant human TGM4 (421-565aa) purified from E. coli
Protein G purified
RUO
Primary
7047
Human
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50
NP_003232
EC 2.3.2.13, fibrinoligase, hTGP, Prostate transglutaminase, Prostate-specific transglutaminase, protein-glutamine gamma-glutamyltransferase 4, TG(P), TGase P, TGase-4, TGPFLJ26776, transglutaminase 4 (prostate), Transglutaminase P, Transglutaminase-4
Mouse
IgG2b
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Anti-human TGM4 mAb, clone 1H1, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TGM4 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only