Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-1 Antibody (4E4), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP141217
Description
TRF-1 Monoclonal specifically detects TRF-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TRF-1 | |
Monoclonal | |
1.0mg/mL | |
Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
TERF1 | |
The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
Primary | |
This antibody will recognize human TRF1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
40000 | |
Unconjugated | |
NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
Mouse | |
70 kDa | |
100 μL | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
7013 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction