Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRF-1, Clone: 8A5, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141219

Catalog No. NBP141219

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TRF-1
8A5
Unconjugated
PBS (pH 7.4) with No Preservative
NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Mouse
IgG
100ul
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary
7013
Human
Western Blot
1.0mg/mL
Western Blot
Purified
TERF1
The exact sequence is proprietary.
Immunogen affinity purified
RUO
Store at -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody will recognize human TRF1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only