Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Troponin I Type 3 (cardiac), Biotin, Clone: 2C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235336B

Catalog No. NBP235336B


Add to cart

Description

Description

Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Troponin I Type 3 (cardiac)
2C9
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
TNNI3
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
PBS with 0.05% Sodium Azide
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Mouse
IgG2b Kappa
0.1 ml
Apoptosis, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers
Primary
7137
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only