Learn More
anti-Troponin I Type 3 (cardiac), Clone: 2C9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235336
 View more versions of this product
Catalog No. NBP235336
Add to cart
Description
Description
Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Troponin I Type 3 (cardiac)
2C9
Unconjugated
PBS with 0.05% Sodium Azide
Purified
TNNI3
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 0.5 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
P19429
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Mouse
IgG2b Kappa
0.1 mg
Apoptosis, Cytoskeleton Markers, Stem Cell Markers
Primary
7137