Learn More
anti-Troponin I Type 3 (cardiac), DyLight 550, Clone: 2C9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP235336R
Catalog No. NBP235336R
Add to cart
Description
Description
Troponin I Type 3 (cardiac) Monoclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Troponin I Type 3 (cardiac)
2C9
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
TNNI3
Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Mouse
IgG2b Kappa
0.1 ml
Apoptosis, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers
Primary
7137