Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPC6 Mouse anti-Human, Clone: 3F2.H10.F2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142738

Catalog No. NBP142738


Add to cart

Description

Description

TRPC6 Monoclonal antibody specifically detects TRPC6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TRPC6
3F2.H10.F2
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
FLJ11098, FSGS2, short transient receptor potential channel 6, transient receptor potential cation channel, subfamily C, member 6, Transient receptor protein 6, TRP-6, TRP6FLJ14863, TrpC6
Mouse
IgG1 Kappa
0.1 mg
Signal Transduction
Primary
7225
Human, Primate
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/ml
Western Blot 1:500-1:2000, ELISA 1:10000-1:50000, Immunohistochemistry 2.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Purified
TRPC6
This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to a region near the carboxy terminus of human TRPC6 protein.
Protein A purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
A BLAST analysis was used to suggest cross-reactivity with TRPC6 from chimpanzee based on 100% identity with the immunizing sequence. Cross-reactivity with TRPC6 from other sources has not been determined.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only