Learn More
anti-u-Plasminogen Activator/Urokinase, Clone: PGM2001, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP142218
Catalog No. NBP142218
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
u-Plasminogen Activator/Urokinase
PGM2001
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Ascites
PLAU
The antibody reacts with the Urokinase antigen.
Unpurified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody binds to human high molecular weight single-chain urokinase type plasminogen activator (HMW-scuPA, 54 kDa), to high molecular weight two-chain urokinase type plasmino¡gen activator (HMW-tcuPA, 52 kDa) and to low molecular weight single-chain urokinase type plasmino¡gen activator (LMW-scuPA, 33 kDa) with high affinity (Kd= 0.1 nM) and does inhibit the ability of urokinase to activate plasminogen. Urokinase reaction with synthetic substrate S2444 is not inhibited. The antibody is highly specific for urokinase, no cross reactions were found with other serine proteases (tissue plasminogen activator, plasmin, trypsin, chymotrypsin, elastase, thrombin). Human plasma or plasma proteins do not interfere with its binding to urokinase.
ELISA, Western Blot
Unconjugated
Ascites with No Preservative
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Mouse
IgG1
1 ml
Cancer
Primary
5328
Human, Rat