Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-UBE2S, Alexa Fluor 405, Clone: 2C12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141126AF405

Catalog No. NBP141126W


Add to cart

Description

Description

UBE2S Monoclonal antibody specifically detects UBE2S in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

UBE2S
2C12
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Q16763
E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S
Mouse
IgG2a Kappa
Protein G purified
RUO
Primary
27338
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
UBE2S
Recombinant human UBE2S (1-222aa) purified from E. coli (NP_055316.2).
24 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only