Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ubiquitin, Clone: 5B9-B3, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP121678

Catalog No. NBP121678

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Ubiquitin
5B9-B3
Unconjugated
PBS (pH 7.4) and 50% Glycerol with 0.09% Sodium Azide
Purified
RPS27A
Native bovine ubiquitin, conjugated to KLH
Protein G purified
RUO
Primary
6233
Bovine
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Western Blot 1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
P0CG47, P0CG53
RPS27A, UBA52, UBB ubiquitin B, UBC
Mouse
IgG2a Kappa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Detects approx 10kDa.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only