Learn More
anti-USF1, Clone: 1264CT170.274.14, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232306
Catalog No. NBP232306
Add to cart
Description
Description
USF1 Monoclonal antibody specifically detects USF1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
USF1
1264CT170.274.14
Unconjugated
PBS with 0.09% Sodium Azide
Purified
USF1
This USF1 antibody is generated from a mice immunized with a recombinant protein between 1-310 amino acids from the Central region of human USF1.
33.538 kDa
400 ul
Transcription Factors and Regulators
Primary
7391
Western Blot
0.5 mg/mL
Western Blot 1:500
P22415
BHLHB11, bHLHb11FCHL1, Class B basic helix-loop-helix protein 11, FCHL, HYPLIP1, Major late transcription factor 1, MLTF, MLTFI, UEF, upstream stimulatory factor 1, upstream transcription factor 1, USF
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human