Learn More
anti-USP25, Clone: 1277CT376.106.171, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232311
Catalog No. NBP232311
Add to cart
Description
Description
USP25 Monoclonal antibody specifically detects USP25 in Human samples. It is validated for Western Blot.
Specifications
Specifications
USP25
1277CT376.106.171
Unconjugated
PBS with 0.09% Sodium Azide
Purified
USP25
This USP25 antibody is generated from a mouse immunized with a recombinant protein from human USP25.
122.218 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
0.5 mg/mL
Western Blot 1:1000
Q9UHP3
Deubiquitinating enzyme 25, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 25, ubiquitin specific peptidase 25, ubiquitin specific protease USP2510USP21ubiquitin specific protease 25, ubiquitin thioesterase 25, Ubiquitin thiolesterase 25, ubiquitin-specific processing protease 25, Ubiquitin-specific-processing protease 25, USP on chromosome 21
Mouse
IgG1 Kappa
Protein G purified
RUO
Primary
29761