Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-VAMP-7, Clone: 158.2, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP107118
Description
VAMP-7 Monoclonal specifically detects VAMP-7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VAMP-7 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:5-1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10 - 1:400 | |
VAMP7 | |
TI-VAMP recombinant protein | |
1 mL | |
Endosome Markers | |
6845 | |
Reconstitute with 1.0 ml distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
158.2 | |
Unconjugated | |
FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Reacts specifically with the 25 kDa protein of human TI-VAMP protein (human, mouse, rat, canine) | |
Human, Mouse, Rat, Canine | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only