Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-VCAM-1/CD106, Biotin, Clone: 1.G11B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128406

Catalog No. NBP128406


Add to cart

Description

Description

VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications

Specifications

VCAM-1/CD106
1.G11B1
Western Blot 1:100-1:2000, Flow Cytometry 10 uL/10^6 cells, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Purified
VCAM1
The immunogen is a peptide made to the amino acid region VCAM1
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human CD106/VCAM-1
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Biotin
PBS with 0.1% Sodium Azide
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Mouse
IgG1 Kappa
1 ml
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
7412
Human, Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only