Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-VDR/NR1I1/Vitamin D Receptor, Clone: 8C356, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP194706

Catalog No. NBP194706


Add to cart

Description

Description

VDR/NR1I1/Vitamin D Receptor Monoclonal antibody specifically detects VDR/NR1I1/Vitamin D Receptor in Human samples. It is validated for Western Blot, Dot Blot.
Specifications

Specifications

VDR/NR1I1/Vitamin D Receptor
8C356
Western Blot 1:100-1:2000, Dot Blot 1:100-1:2000
Purified
VDR
Partial recombinant sequence from human Vitamin D Receptor
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Dot Blot, Western Blot
Unconjugated
PBS (pH 7.4) and 1.0% BSA with 0.05% Sodium Azide
NR1I1Nuclear receptor subfamily 1 group I member 11,25-dihydroxyvitamin D3 receptor, vitamin D1,25- dihydroxyvitamin D3 receptor, vitamin D3 receptor
Mouse
IgG1
0.05 mg
Signal Transduction
Primary
7421
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only