Learn More
Mouse VEGFR1/Flt-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 27 publications
Supplier:  R&D Systems AF471
Catalog No. AF471
Add to Cart
This item is not returnable. View return policy
Description
Description
VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
VEGFR1/Flt-1
Polyclonal
Unconjugated
P35969
FLT1
Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1/Flt-1, Ser27-Glu759, Accession # P35969
100 μg
Primary
Detects mouse VEGFR1/Flt-1 in ELISAs and Western blots.
Mouse
IgG
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Neutralization, CyTOF, ELISA
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Neutralization 2-8 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1
Goat
Affinity Purified
RUO
2321
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions