Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Human, Mouse (Negative), Alexa Fluor 647, Clone: 2D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192687AF647

Catalog No. NBP192687Y


Add to cart

Description

Description

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay.
Specifications

Specifications

Vimentin
2D1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
P08670
FLJ36605, vimentin
Mouse
IgG2a
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Rat and Mouse and likely all other Mammals.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
VIM
Recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670]
50 kDa
0.1 ml
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
Primary
7431
Human, Mouse (Negative), Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only