Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Vitronectin, Clone: BE10, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142299

Catalog No. NBP142299

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Vitronectin
BE10
Unconjugated
PBS solution containing 1.0% w/v BSA with 0.1% Sodium Azide
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Mouse
IgG1
0.05 mg
Immunology, Innate Immunity
Primary
7448
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
VTN
The antibody reacts with the Vitronectin antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The antibody is specific for vitronectin in human tissues. No cross reaction with other matrix molecules. Vitronectin, also known as serum spreading factor, S-protein and epibolin, is a glycoprotein present both in human plasma and serum. Vitronectin has been shown to modulate blood coagulation and complement-induced cytolysis. It is also variably present in diverse loose connective tissues, often in co-localization elastic fibrils. Clone BE10 recognizes vitronectin specifically and can be used in immunohistochemical stainings, immunoblottings and other immunoassays for cellular and tumor biology. Monoclonal antibody to vitronectin is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells. Purified vitronectin from human plasma was used as an immunogen.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only