Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Vitronectin, Clone: BV1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142380

Catalog No. NBP142380


Add to cart

Description

Description

Vitronectin Monoclonal antibody specifically detects Vitronectin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

Vitronectin
BV1
Unconjugated
PBS, 0.1% BSA with 0.02% Sodium Azide
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Mouse
IgG1
0.2 ml
Immunology, Innate Immunity
Primary
7448
Human
Flow Cytometry, Immunoprecipitation, Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:10-1:500
Purified
VTN
The antibody reacts with the Vitronectin antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This monoclonal antibody (subclass IgG1) binds to human vitronectin. It binds to soluble vitronectin as well as to membrane bound vitronectin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only