Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VLDLR Antibody (6A6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178162UV
Description
VLDLR Monoclonal specifically detects VLDLR in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VLDLR | |
Monoclonal | |
DyLight 350 | |
P98155 | |
VLDLR | |
Synthetic peptide corresponding to the C-terminus of human VLDL Receptor [Swiss-Prot# P98155] | |
0.1 mL | |
Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
7436 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
6A6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction