Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-1 Mouse anti-Human, Clone: 13F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142739

Catalog No. NBP142739


Add to cart

Description

Description

Wnt-1 Monoclonal antibody specifically detects Wnt-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Wnt-1
13F9
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
INT1Proto-oncogene Int-1 homolog, proto-oncogene Wnt-1, wingless-type MMTV integration site family, member 1, wingless-type MMTV integration site family, member 1 (oncogene INT1)
Mouse
IgG1 Kappa
0.1 mg
Cellular Markers, Wnt Signaling Pathway
Primary
7471
ELISA, Western Blot
1 mg/ml
Western Blot 1:175-1:250, ELISA 1:4000-1:20000
Purified
WNT1
Synthetic peptide corresponding to an internal region of human Wnt1 protein.. Uniprot ID P04628
Protein A purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bovine, Canine, Human, Monkey, Mouse, Opossum, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only